5,6,7,8-Tetrahydroimidazo[1,5-a]pyrazine 97%

Supplier: Ambeed
Warning

A299726-50MG A299726-100MG A299726-250MG
103814-240EA 99 USD
103814-240 103835-390 76802-516 77601-346 77668-336
5,6,7,8-Tetrahydroimidazo[1,5-a]pyrazine 97%
5,6,7,8-Tetrahydroimidazo[1,5-a]pyrazine

Precaución: for Research Use Only

Formula: C₆H₉N₃
MW: 123.16 g/mol
Boiling Pt: 340 °C (760 mm Hg)
Flash Pt: 169.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09263955
CAS Number: 297172-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR