5,6,7,8-Tetrahydroimidazo[1,2-a]pyrazine hydrochloride 95%

Supplier: Ambeed
Warning

A162708-100MG A162708-250MG A162708-1G
103805-652EA 29 USD
103805-652 103843-902 77028-672 77028-674 77300-602
5,6,7,8-Tetrahydroimidazo[1,2-a]pyrazine hydrochloride 95%
5,6,7,8-Tetrahydroimidazo[1,2-a]pyrazine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₀ClN₃
MW: 159.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878589
CAS Number: 1187830-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR