5,6,7,8-Tetrahydro-1,7-naphthyridine dihydrochloride 97%

Supplier: Ambeed
Warning

A468199-100MG A468199-250MG A468199-5G A468199-1G
103828-000EA 197 USD
103828-000 103839-074 76818-490 76818-492 76842-980 77307-458
5,6,7,8-Tetrahydro-1,7-naphthyridine dihydrochloride 97%
5,6,7,8-Tetrahydro-1,7-naphthyridine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂Cl₂N₂
MW: 207.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09056821
CAS Number: 351038-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR