5,6,7,8-Tetrahydro[1,2,4]triazolo[1,5-a]pyrazine 97%

Supplier: Ambeed
Warning

A118468-100MG A118468-10G A118468-250MG A118468-1G A118468-5G
103799-064EA 56 USD
103799-064 77094-350 77094-352 77094-354 77311-734 77311-736
5,6,7,8-Tetrahydro[1,2,4]triazolo[1,5-a]pyrazine 97%
5,6,7,8-Tetrahydro[1,2,4]triazolo[1,5-a]pyrazine

Precaución: for Research Use Only

Formula: C₅H₈N₄
Flash Pt: 147.187 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09835083
CAS Number: 233278-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR