4,4'-Oxydiphthalic anhydride 97%

Supplier: Ambeed
Warning

Synonyms: 5,5'-Oxybis(isobenzofuran-1,3-dione)

A102998-100G A102998-10G A102998-25G
103822-164EA 6 USD
103822-164 77025-226 77025-228 77025-230
4,4'-Oxydiphthalic anhydride 97%
4,4'-Oxydiphthalic anhydride

Precaución: for Research Use Only

Formula: C₁₆H₆O₇
MW: 310.22 g/mol
Boiling Pt: 577.7 °C (760 mm Hg)
Melting Pt: 225…229 °C
Storage Temperature: Ambient
MDL Number: MFCD00039144
CAS Number: 1823-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR