5,5-Dimethylpyrrolidin-2-one 98%

Supplier: Ambeed
Warning

A110619-100MG A110619-250MG A110619-1G A110619-5G A110619-10G
103797-694EA 23 USD
103797-694 103823-502 77025-754 77025-756 77310-302 77310-304 77871-449
5,5-Dimethylpyrrolidin-2-one 98%
5,5-Dimethylpyrrolidin-2-one

Precaución: for Research Use Only

Formula: C₆H₁₁NO
Boiling Pt: 229.2 °C (760 mm Hg)
Melting Pt: 39…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00128876
CAS Number: 5165-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR