5,5-Dimethylhydantoin 97%

Supplier: Ambeed
Warning

Synonyms: 5,5-Dimethylimidazolidine-2,4-dione

A448794-25G A448794-100G A448794-500G
103810-348EA 35 USD
103810-348 76842-660 76877-480 76877-482
5,5-Dimethylhydantoin 97%
5,5-Dimethylhydantoin

Precaución: for Research Use Only

Formula: C₅H₈N₂O₂
MW: 128.13 g/mol
Melting Pt: 175…180 °C
Flash Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00005266
CAS Number: 77-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR