5,5'-Dimethyl-2,2'-bipyridyl 98%

Supplier: Ambeed
Warning

A167981-100G A167981-10G A167981-1G A167981-25G A167981-5G
103806-234EA 14 USD
103806-234 103844-492 76998-098 76998-100 77081-526 77081-528 77081-530
5,5'-Dimethyl-2,2'-bipyridyl 98%
5,5'-Dimethyl-2,2'-bipyridyl

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂
Boiling Pt: 315.7±37.0 °C (760 mm Hg)
Melting Pt: 114…117 °C
Flash Pt: 119.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01740554
CAS Number: 1762-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR