5,5'-Dibromo-2,2'-bithiophene 98%

Supplier: Ambeed
Warning

A103154-100G A103154-10G A103154-25G A103154-500G A103154-1G A103154-5G
103796-492EA 6 USD
103796-492 103822-196 76765-282 76765-284 76765-286 76765-288 77283-050 77309-068
5,5'-Dibromo-2,2'-bithiophene 98%
5,5'-Dibromo-2,2'-bithiophene

Precaución: for Research Use Only

Formula: C₈H₄Br₂S₂
MW: 324.05 g/mol
Boiling Pt: 318.6 °C (760 mm Hg)
Melting Pt: 144…146 °C
Flash Pt: 146.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00219110
CAS Number: 4805-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR