5,5'-Dibromo-2,2'-bipyridyl 97%

Supplier: Ambeed
Warning

A211805-100MG A211805-10G A211805-250MG A211805-1G A211805-25G
103818-142EA 29 USD
103818-142 103848-836 76814-564 76814-566 76814-568 77087-264 77883-507
5,5'-Dibromo-2,2'-bipyridyl 97%
5,5'-Dibromo-2,2'-bipyridyl

Precaución: for Research Use Only

Formula: C₁₀H₆Br₂N₂
Boiling Pt: 357.7 °C (760 mm Hg)
Flash Pt: 170.1 °C
Storage Temperature: Ambient
MDL Number: MFCD06657883
CAS Number: 15862-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR