Bis(neopentyl glycolato)diboron 98%

Supplier: Ambeed
Warning

Synonyms: 5,5,5',5'-Tetramethyl-2,2'-bi(1,3,2-dioxaborinane), 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane

A206770-100G A206770-10G A206770-1G A206770-25G A206770-5G A206770-500G
103848-408EA 6 USD
103848-408 76691-328 76691-330 76691-332 77086-664 77086-666 77550-324
Bis(neopentyl glycolato)diboron 98%
Bis(neopentyl glycolato)diboron

Precaución: for Research Use Only

Formula: C₁₀H₂₀B₂O₄
MW: 225.89 g/mol
Boiling Pt: 214.3 °C (760 mm Hg)
Melting Pt: 180.5…184.5 °C
Flash Pt: 83.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093062
CAS Number: 201733-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR