Honokiol 98%

Supplier: Ambeed
Danger

A146214-10MG A146214-250MG A146214-50MG A146214-25G A146214-1G A146214-5G
103803-644EA 11 USD
103803-644 103841-730 77285-936 77285-938 77285-940 77299-504 77317-042 77317-044
Honokiol 98%
Honokiol

Precaución: for Research Use Only

Formula: C₁₈H₁₈O₂
Boiling Pt: 400.1 °C (760 mm Hg)
Flash Pt: 184 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00016674
CAS Number: 35354-74-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR