5,10,15,20-Tetraphenyl-21H,23H-porphine copper(II) 98%

Supplier: Ambeed
Warning

Synonyms: Copper TPP, Copper(II) Tetraphenylporphyrin, Copper meso-tetraphenylporphine, Copper(II) TPP, 5,10,15,20-Tetraphenyl-21H,23H-porphine Copper(II)

A105701-100MG A105701-1G A105701-5G A105701-250MG
103796-926EA 71 USD
103796-926 103822-664 76687-246 77283-386 77283-388 77570-332
5,10,15,20-Tetraphenyl-21H,23H-porphine copper(II) 98%
5,10,15,20-Tetraphenyl-21H,23H-porphine copper(II)

Precaución: for Research Use Only

Formula: C₄₄H₂₈CuN₄
MW: 676.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00148862
CAS Number: 14172-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR