5-Trimethylsilyl-4-pentyn-1-ol 95%

Supplier: Ambeed
Warning

Synonyms: 5-(Trimethylsilyl)pent-4-yn-1-ol

A121450-10G A121450-5G A121450-25G
103800-078EA 17 USD
103800-078 76766-510 77297-832 77596-346
5-Trimethylsilyl-4-pentyn-1-ol 95%
5-Trimethylsilyl-4-pentyn-1-ol

Precaución: for Research Use Only

Formula: C₈H₁₆OSi
MW: 156.30 g/mol
Boiling Pt: 189.6 °C (760 mm Hg)
Flash Pt: 68.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00190229
CAS Number: 13224-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR