5-(Trifluoromethyl)thiazol-2-amine 95%

Supplier: Ambeed
Warning

A187692-100MG A187692-250MG A187692-5G A187692-1G
103816-038EA 145 USD
103816-038 77029-700 77029-702 77029-704 77084-344
5-(Trifluoromethyl)thiazol-2-amine 95%
5-(Trifluoromethyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₄H₃F₃N₂S
MW: 168.14 g/mol
Boiling Pt: 214 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11846867
CAS Number: 169260-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR