5-(Trifluoromethyl)pyridine-2,3-diamine 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-Diamino-5-(trifluoromethyl)pyridine

A180845-5G A180845-100MG A180845-250MG
103807-568EA 107 USD
103807-568 103845-930 77083-440 77248-264 77248-266
5-(Trifluoromethyl)pyridine-2,3-diamine 97%
5-(Trifluoromethyl)pyridine-2,3-diamine

Precaución: for Research Use Only

Formula: C₆H₆F₃N₃
MW: 177.127 g/mol
Boiling Pt: 296.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD10696252
CAS Number: 107867-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR