2-Hydroxy-5-(trifluoromethyl)pyridine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-(Trifluoromethyl)pyridin-2-ol
A157588-100G
A157588-10G
A157588-500G
A157588-25G
A157588-5G
103843-218EA
5
USD
103843-218
77096-046
77096-048
77096-050
77319-050
77319-052
2-Hydroxy-5-(trifluoromethyl)pyridine 97%
2-Hydroxy-5-(trifluoromethyl)pyridine
Precaución: for Research Use Only
Formula:
C₆H₄F₃NO MW: 163.1 Boiling Pt: 286.8 °C (760 mm Hg) Melting Pt: 145…149 °C Flash Pt: 127.262 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042315 CAS Number: 33252-63-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...