5-(Trifluoromethyl)pyridazin-3-amine 95%
Supplier: Ambeed
|
Warning
|
A122818-100MG
A122818-250MG
103800-288EA
387
USD
103800-288
77161-414
77161-416
5-(Trifluoromethyl)pyridazin-3-amine 95%
5-(Trifluoromethyl)pyridazin-3-amine
Precaución: for Research Use Only
Formula:
C₅H₄F₃N₃ MW: 163.1 g/mol Boiling Pt: 294.8±40.0 °C (760 mm Hg) Flash Pt: 132.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD20726836 CAS Number: 1211591-88-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...