5-(Trifluoromethyl)picolinonitrile 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Cyano-5-(trifluoromethyl)pyridine
A118251-100G
A118251-10G
A118251-25G
A118251-1G
A118251-5G
103799-018EA
5
USD
103799-018
103824-906
76810-302
76810-304
76810-306
77311-698
77311-700
5-(Trifluoromethyl)picolinonitrile 95%
5-(Trifluoromethyl)picolinonitrile
Precaución: for Research Use Only
Formula:
C₇H₃F₃N₂ MW: 172.11 g/mol Boiling Pt: 232.3 °C (760 mm Hg) Flash Pt: 94.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01001119 CAS Number: 95727-86-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...