5-(Trifluoromethyl)picolinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-(Trifluoromethyl)pyridine-2-carboxylic acid

A207511-100G A207511-10G A207511-250MG A207511-1G A207511-25G A207511-5G
103817-802EA 6 USD
103817-802 103848-484 76770-356 76770-358 76770-360 77086-762 77086-764 77303-038
5-(Trifluoromethyl)picolinic acid 97%
5-(Trifluoromethyl)picolinic acid

Precaución: for Research Use Only

Formula: C₇H₄F₃NO₂
MW: 191.11 g/mol
Melting Pt: 133…137 °C
Flash Pt: 119.353 °C
Storage Temperature: Ambient
MDL Number: MFCD04113632
CAS Number: 80194-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR