5-(Trifluoromethyl)pyridine-2-carboxaldehyde 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-(Trifluoromethyl)picolinaldehyde
A427709-1G
A427709-25G
A427709-100MG
A427709-10G
A427709-250MG
A427709-100G
103827-488EA
96
USD
103827-488
103838-542
76842-200
76842-202
77016-522
77016-524
77016-526
77890-875
5-(Trifluoromethyl)pyridine-2-carboxaldehyde 95%
5-(Trifluoromethyl)pyridine-2-carboxaldehyde
Precaución: for Research Use Only
Formula:
C₇H₄F₃NO Melting Pt: 31…33 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07375388 CAS Number: 31224-82-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...