5-(Trifluoromethyl)indole 97%

Supplier: Ambeed
Warning

A201955-1G A201955-25G A201955-5G A201955-10G A201955-250MG
103817-340EA 42 USD
103817-340 103847-978 77086-068 77086-070 77086-072 77274-868 77274-870
5-(Trifluoromethyl)indole 97%
5-(Trifluoromethyl)indole

Precaución: for Research Use Only

Formula: C₉H₆F₃N
MW: 185.15 g/mol
Melting Pt: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD03095341
CAS Number: 100846-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR