5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-(Trifluoromethyl)-5H-dibenzo[b,d]thiophen-5-ium trifluoromethanesulphonate
A218833-10G
A218833-250MG
A218833-1G
A218833-5G
A218833-25G
103818-752EA
63
USD
103818-752
103849-622
76721-900
76721-902
77088-160
77088-162
77303-516
5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate 97%
5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate
Precaución: for Research Use Only
Formula:
C₁₄H₈F₆O₃S₂ MW: 402.325 g/mol Melting Pt: 150 °C Flash Pt: 81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236132 CAS Number: 129946-88-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...