5-(Trifluoromethoxy)pyridin-2-amine 96%

Supplier: Ambeed
Warning

A144171-100MG A144171-250MG
103803-366EA 544 USD
103803-366 103841-444 76871-350 77611-322
5-(Trifluoromethoxy)pyridin-2-amine 96%
5-(Trifluoromethoxy)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₅F₃N₂O
MW: 178.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19690149
CAS Number: 1221171-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR