5-(Trifluoromethoxy)isatin 98%

Supplier: Ambeed
Warning

A143224-10G A143224-250MG A143224-1G
103803-250EA 17 USD
103803-250 76996-938 76996-940 77299-330
5-(Trifluoromethoxy)isatin 98%
5-(Trifluoromethoxy)isatin

Precaución: for Research Use Only

Formula: C₉H₄F₃NO₃
Melting Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00192524
CAS Number: 169037-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR