5-(tert-Butyl)isoxazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A169781-100MG A169781-10G A169781-1G A169781-5G A169781-250MG
103806-408EA 74 USD
103806-408 103844-672 76784-160 76784-162 77081-774 77081-776 77667-368
5-(tert-Butyl)isoxazole-3-carboxylic acid 95%
5-(tert-Butyl)isoxazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₁NO₃
MW: 169.1800 g/mol
Flash Pt: 137.328 °C
Storage Temperature: Freezer
MDL Number: MFCD07377301
CAS Number: 90607-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR