5-tert-Butylsalicylaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 5-(tert-Butyl)-2-hydroxybenzaldehyde, 5-tert-Butyl-2-hydroxybenzaldehyde

A214267-1G A214267-5G A214267-10G
103818-382EA 7 USD
103818-382 103849-088 77087-598 77303-336 77568-064
5-tert-Butylsalicylaldehyde 98%
5-tert-Butylsalicylaldehyde

Precaución: For research use only.

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 252 °C (760 mm Hg)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD01321335
CAS Number: 2725-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR