3-Pyridin-3-yl-1H-pyrazol-5-amine 95%
Supplier: Ambeed
|
Warning
|
A177379-250MG
A177379-5G
A177379-100MG
103807-230EA
210
USD
103807-230
103845-558
77011-822
77301-504
77890-132
3-Pyridin-3-yl-1H-pyrazol-5-amine 95%
3-Pyridin-3-yl-1H-pyrazol-5-amine
Precaución: for Research Use Only
Formula:
C₈H₈N₄ Boiling Pt: 486.9 °C (760 mm Hg) Flash Pt: 280.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD06738753 CAS Number: 149246-87-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...