2-Amino-5-(4-methylphenyl)-1,3,4-thiadiazole 95%

Supplier: Ambeed
Warning

Synonyms: 4-(5-Amino-1,3,4-thiadiazol-2-yl)toluene, 5-(4-Methylphenyl)-1,3,4-thiadiazol-2-amine, 5-(p-Tolyl)-1,3,4-thiadiazol-2-amine

A150550-10G A150550-250MG A150550-1G A150550-5G
103804-180EA 54 USD
103804-180 103842-316 77247-226 77247-228 77317-836 77317-838
2-Amino-5-(4-methylphenyl)-1,3,4-thiadiazole 95%
2-Amino-5-(4-methylphenyl)-1,3,4-thiadiazole

Precaución: for Research Use Only

Formula: C₉H₉N₃S
MW: 191.253 g/mol
Boiling Pt: 364.9 °C (760 mm Hg)
Melting Pt: 218…222 °C
Flash Pt: 174.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00469688
CAS Number: 26907-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR