5-(Piperazin-1-yl)benzofuran-2-carboxamide 97%

Supplier: Ambeed
Warning

A307370-1G A307370-25G A307370-5G A307370-10G A307370-250MG
103814-500EA 12 USD
103814-500 103835-676 76838-414 76838-416 76838-418 76875-712 76875-714
5-(Piperazin-1-yl)benzofuran-2-carboxamide 97%
5-(Piperazin-1-yl)benzofuran-2-carboxamide

Precaución: for Research Use Only

Formula: C₁₃H₁₅N₃O₂
Boiling Pt: 505.319 °C (760 mm Hg)
Melting Pt: 256…260 °C
Flash Pt: 259.408 °C
Storage Temperature: Ambient
MDL Number: MFCD20486139
CAS Number: 183288-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR