5-(Methoxymethylene)-2,2-dimethyl-1,3-dioxane-4,6-dione 97% (by HPLC)
Supplier: Ambeed
|
Warning
|
A403273-10G
A403273-25G
A403273-1G
A403273-5G
A403273-100G
103827-090EA
5
USD
103827-090
103838-130
76773-742
76773-744
76841-602
76841-604
77878-144
5-(Methoxymethylene)-2,2-dimethyl-1,3-dioxane-4,6-dione 97% (by HPLC)
5-(Methoxymethylene)-2,2-dimethyl-1,3-dioxane-4,6-dione
Precaución: for Research Use Only
Formula:
C₈H₁₀O₅ Boiling Pt: 390 °C (760 mm Hg) Melting Pt: 132…134 °C (760 mm Hg) Flash Pt: 178.7 °C Storage Temperature: Freezer |
MDL Number:
MFCD00599240 CAS Number: 15568-85-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...