Thiophene-2,5-dicarboxylic acid monomethyl ester 98%

Supplier: Ambeed
Warning

Synonyms: 5-(Methoxycarbonyl)thiophene-2-carboxylic acid

A114742-250MG A114742-5G
103798-462EA 19 USD
103798-462 103824-304 76766-076 77311-126
Thiophene-2,5-dicarboxylic acid monomethyl ester 98%
Thiophene-2,5-dicarboxylic acid monomethyl ester

Precaución: for Research Use Only

Formula: C₇H₆O₄S
MW: 186.19 g/mol
Melting Pt: 193…197 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01203161
CAS Number: 50340-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR