5-(Methoxycarbonyl)-2-pyridine carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-(Methoxycarbonyl)picolinic acid

A125833-100G A125833-10G A125833-500G A125833-1G A125833-25G A125833-5G
103800-730EA 9 USD
103800-730 76996-088 76996-090 76996-092 77298-092 77298-094 77312-878
5-(Methoxycarbonyl)-2-pyridine carboxylic acid 97%
5-(Methoxycarbonyl)-2-pyridine carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 347.9 °C (760 mm Hg)
Melting Pt: 187 °C
Flash Pt: 164.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00179343
CAS Number: 17874-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR