5-(Hydroxymethyl)picolinic acid 95%

Supplier: Ambeed
Warning

A120787-100MG A120787-250MG A120787-1G
103799-994EA 72 USD
103799-994 76983-248 76983-250 77297-788
5-(Hydroxymethyl)picolinic acid 95%
5-(Hydroxymethyl)picolinic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₃
MW: 153.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11520014
CAS Number: 39977-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR