5-(2-Furyl)isoxazole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-(Furan-2-yl)isoxazole-3-carboxylic acid

A223668-1G A223668-10G A223668-250MG
103819-056EA 57 USD
103819-056 103849-936 77088-560 77237-688 77237-690
5-(2-Furyl)isoxazole-3-carboxylic acid 97%
5-(2-Furyl)isoxazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅NO₄
MW: 179.13 g/mol
Boiling Pt: 418.4 °C (760 mm Hg)
Melting Pt: 151 °C
Storage Temperature: Freezer
MDL Number: MFCD04113954
CAS Number: 98434-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR