5-(Difluoromethyl)pyrazine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A402156-1G A402156-100MG
103827-072EA 29 USD
103827-072 76841-568 76876-912
5-(Difluoromethyl)pyrazine-2-carboxylic acid 95%
5-(Difluoromethyl)pyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₄F₂N₂O₂
MW: 174.105 g/mol
Boiling Pt: 319.8 °C (760 mm Hg)
Flash Pt: 147.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD20640530
CAS Number: 1174321-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR