5-(Difluoromethoxy)-2-mercaptobenzimidazole 97%

Supplier: Ambeed
Warning

Synonyms: 5-(Difluoromethoxy)-2-benzimidazolethiol, 5-Difluoromethoxy-2-mercaptobenzimidazole, 5-(Difluoromethoxy)-1H-benzo[d]imidazole-2-thiol, 5-(Difluoromethoxy)-2-mercapto-1H-benzimidazole

A223953-10G A223953-500G A223953-100G
103849-962EA 5 USD
103849-962 77237-700 77237-702 77746-321
5-(Difluoromethoxy)-2-mercaptobenzimidazole 97%
5-(Difluoromethoxy)-2-mercaptobenzimidazole

Precaución: for Research Use Only

Formula: C₈H₆F₂N₂OS
MW: 216.21 g/mol
Melting Pt: 239…243 °C
Storage Temperature: Freezer
MDL Number: MFCD00467504
CAS Number: 97963-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR