5-(Cyanomethyl)-2-fluorobenzonitrile 95%

Supplier: Ambeed
Warning

A135748-100MG A135748-250MG A135748-1G
103802-214EA 55 USD
103802-214 77109-506 77109-508 77298-816
5-(Cyanomethyl)-2-fluorobenzonitrile 95%
5-(Cyanomethyl)-2-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₉H₅FN₂
MW: 160.1500 g/mol
Boiling Pt: 302 °C (760 mm Hg)
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD03094342
CAS Number: 519059-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR