5-(Chloromethyl)-4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole 97%

Supplier: Ambeed
Danger

Synonyms: 5-(Chloromethyl)-4-methyl-2-(4-(trifluoromethyl)phenyl)thiazole

A289827-1G A289827-5G
103813-848EA 12 USD
103813-848 103834-988 76837-570 76837-572
5-(Chloromethyl)-4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole 97%
5-(Chloromethyl)-4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole

Precaución: for Research Use Only

Formula: C₁₂H₉ClF₃NS
MW: 291.72 g/mol
Melting Pt: 60…62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03791173
CAS Number: 317318-97-1
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR