5-(Chloromethyl)-2-methylpyridine hydrochloride 97%

Supplier: Ambeed
Warning

A181429-100MG A181429-250MG A181429-1G
103807-604EA 68 USD
103807-604 103845-976 76985-774 76985-776 77301-718
5-(Chloromethyl)-2-methylpyridine hydrochloride 97%
5-(Chloromethyl)-2-methylpyridine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉Cl₂N
MW: 178.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11111128
CAS Number: 106651-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR