5-(Chloromethyl)-2-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Danger

A503585-100MG A503585-10G A503585-250MG A503585-1G A503585-25G A503585-5G
103828-314EA 23 USD
103828-314 103839-406 76726-042 76726-044 76726-046 76843-378 76843-380 76843-382
5-(Chloromethyl)-2-(trifluoromethyl)pyridine 98%
5-(Chloromethyl)-2-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₅ClF₃N
Boiling Pt: 204.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01862650
CAS Number: 386715-33-9
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR