5-(Bromomethyl)pyrimidine hydrobromide 95%

Supplier: Ambeed
Danger

A470510-250MG A470510-1G
103828-022EA 223 USD
103828-022 103839-096 77016-918 77307-476
5-(Bromomethyl)pyrimidine hydrobromide 95%
5-(Bromomethyl)pyrimidine hydrobromide

Precaución: for Research Use Only

Formula: C₅H₆Br₂N₂
MW: 253.92 g/mol
Storage Temperature: Freezer
MDL Number: MFCD24386894
CAS Number: 93224-07-8
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR