5-(Bromomethyl)-2-methylpyridine hydrobromide 95%

Supplier: Ambeed
Danger

A105332-10G A105332-250MG A105332-1G
103796-872EA 63 USD
103796-872 103822-602 77008-566 77008-568 77309-472
5-(Bromomethyl)-2-methylpyridine hydrobromide 95%
5-(Bromomethyl)-2-methylpyridine hydrobromide

Precaución: for Research Use Only

Formula: C₇H₉Br₂N
Storage Temperature: Refrigerator
MDL Number: MFCD12031798
CAS Number: 718608-10-7
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR