3,4,5-Trifluorobenzyl bromide 98%

Supplier: Ambeed
Danger

Synonyms: 5-(Bromomethyl)-1,2,3-trifluorobenzene, α-Bromo-3,4,5-trifluorotoluene

A798961-5G A798961-10G A798961-1G
103841-054EA 30 USD
103841-054 76845-298 77105-584 77308-352
3,4,5-Trifluorobenzyl bromide 98%
3,4,5-Trifluorobenzyl bromide

Precaución: for Research Use Only

Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Boiling Pt: 188.4 °C (760 mm Hg)
Flash Pt: 65.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00083528
CAS Number: 220141-72-0
UN: 1760
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR