5-(Benzyloxy)picolinonitrile 95%

Supplier: Ambeed
Warning

A104515-250MG A104515-1G A104515-5G
103796-726EA 56 USD
103796-726 103822-444 76821-444 77309-308 77309-310
5-(Benzyloxy)picolinonitrile 95%
5-(Benzyloxy)picolinonitrile

Precaución: for Research Use Only

Formula: C₁₃H₁₀N₂O
MW: 210.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11977436
CAS Number: 78760-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR