5-(Benzyloxy)-2-chloropyrimidine 97%

Supplier: Ambeed
Warning

A149814-100MG A149814-250MG A149814-1G A149814-25G A149814-10G
103804-086EA 20 USD
103804-086 103842-214 76997-290 76997-292 77317-724 77317-726 77671-108
5-(Benzyloxy)-2-chloropyrimidine 97%
5-(Benzyloxy)-2-chloropyrimidine

Precaución: for Research Use Only

Formula: C₁₁H₉ClN₂O
Boiling Pt: 387 °C (760 mm Hg)
Flash Pt: 187.9 °C
Storage Temperature: Ambient
MDL Number: MFCD09744017
CAS Number: 138274-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR