5-(Benzyloxy)-2-(4-(benzyloxy)phenyl)-3-methyl-1H-indole 97%

Supplier: Ambeed
Warning

A100829-100G A100829-250MG A100829-25G A100829-1G A100829-5G
103795-646EA 8 USD
103795-646 103821-342 77270-170 77270-172 77270-174 77282-580 77282-582
5-(Benzyloxy)-2-(4-(benzyloxy)phenyl)-3-methyl-1H-indole 97%
5-(Benzyloxy)-2-(4-(benzyloxy)phenyl)-3-methyl-1H-indole

Precaución: for Research Use Only

Formula: C₂₉H₂₅NO₂
Boiling Pt: 620.5 °C (760 mm Hg)
Flash Pt: 213.9 °C
Storage Temperature: Ambient
MDL Number: MFCD04004106
CAS Number: 198479-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR