5-(Benzyloxy)-2-(2,5-dimethyl-1H-pyrrol-1-yl)pyridine 97%

Supplier: Ambeed
Warning

A101030-100MG A101030-250MG A101030-1G A101030-5G
103795-766EA 46 USD
103795-766 103821-458 77160-184 77160-186 77296-960 77296-962
5-(Benzyloxy)-2-(2,5-dimethyl-1H-pyrrol-1-yl)pyridine 97%
5-(Benzyloxy)-2-(2,5-dimethyl-1H-pyrrol-1-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₈H₁₈N₂O
MW: 278.35 g/mol
Boiling Pt: 445.3±45.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD19689570
CAS Number: 1083329-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR