5-(4-Nitrophenyl)-1H-tetrazole 97%

Supplier: Ambeed
Danger

A334869-100MG A334869-1G A334869-5G
103825-668EA 65 USD
103825-668 103836-638 77141-988 77306-472 77306-474
5-(4-Nitrophenyl)-1H-tetrazole 97%
5-(4-Nitrophenyl)-1H-tetrazole

Precaución: for Research Use Only

Formula: C₇H₅N₅O₂
MW: 191.15 g/mol
Boiling Pt: 435.3 °C (760 mm Hg)
Melting Pt: 223 °C
Flash Pt: 217.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00068729
CAS Number: 16687-60-8
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR