5-(4-Methylpiperazino)-2-nitroaniline 95%

Supplier: Ambeed
Warning

Synonyms: 5-(4-Methylpiperazin-1-yl)-2-nitroaniline

A238374-10G A238374-250MG A238374-1G A238374-25G
103811-224EA 13 USD
103811-224 103851-004 77000-348 77000-350 77090-118 77304-106
5-(4-Methylpiperazino)-2-nitroaniline 95%
5-(4-Methylpiperazino)-2-nitroaniline

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₄O₂
MW: 236.28 g/mol
Boiling Pt: 444.2 °C (760 mm Hg)
Melting Pt: 152…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00545964
CAS Number: 23491-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR